This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02289477_g1
See other ROPGEF12 GE Assays ›
Gene Symbol:
ROPGEF12
Gene Name:
RHO guanyl-nucleotide exchange factor 12
Gene Aliases:
AT1G79860, ATROPGEF12, F19K16.18, F19K16_18, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, MATERNAL EFFECT EMBRYO ARREST 64, MEE64, RHO guanyl-nucleotide exchange factor 12, ROP (rho of plants) guanine nucleotide exchange factor 12
Chromosome Location:
Chr.1: 30041989 - 30044297 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02289477_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_106635.4 NP_178104.1 1 - 2 592 56
GenBank mRNA BT033042.1 - 1 - 2 387 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

embryo development ending in seed dormancy
pollen tube growth
regulation of pollen tube growth
Rho guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0