Assay Details
| Gene Symbol: SGT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117197.4 | NP_192865.1 | 2 - 3 | 426 | 82 | |
| GenBank mRNA | AF370229.1 | - | 2 - 3 | 369 | 82 | |
| AF428340.1 | - | 2 - 3 | 364 | 82 | ||
| AF439976.1 | - | 2 - 3 | 233 | 82 | ||
| AJ318019.1 | - | 2 - 3 | 272 | 82 | ||
| AY085464.1 | - | 2 - 3 | 289 | 82 | ||
| AY150487.1 | - | 2 - 3 | 233 | 82 | ||
More Information
Set Membership: |
|

SGT1B
Genomic Map