This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02299878_m1
See other AGL14 GE Assays ›
Gene Symbol:
AGL14
Gene Name:
AGAMOUS-like 14
Gene Aliases:
AGAMOUS-like 14, AT4G11880, T26M18.90, T26M18_90, XAANTAL2, XAL2
Chromosome Location:
Chr.4: 7142922 - 7147391 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02299878_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGL14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001340739.1 NP_001319907.1 2 - 3 496 119
NM_001340740.1 NP_001329521.1 2 - 3 496 119
NM_001340741.1 NP_001329520.1 2 - 3 496 119
NM_117258.5 NP_192925.1 2 - 3 303 119
GenBank mRNA AK230464.1 - 2 - 3 304 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

MADS box transcription factor

Gene Ontology Categories:

Function(s) Process(es)

maintenance of floral meristem identity
regulation of root meristem growth
vegetative to reproductive phase transition of meristem
positive regulation of transcription from RNA polymerase II promoter
regulation of auxin polar transport
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0