This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02300127_g1
See other AT4G18870 GE Assays ›
Gene Symbol:
AT4G18870
Gene Name:
E2F/DP family winged-helix DNA-binding domain-containing protein
Gene Aliases:
AT4G18870, F13C5.40, F13C5_40
Chromosome Location:
Chr.4: 10346168 - 10347316 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02300127_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT4G18870
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_118003.3 NP_193622.1 2 - 3 616 123
GenBank mRNA AB493684.1 - 2 - 3 615 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cellular response to heat
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0