This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02311365_g1
See other AT5G35380 GE Assays ›
Gene Symbol:
AT5G35380
Gene Name:
kinase with adenine nucleotide alpha hydrolases-like domain-containing protein
Gene Aliases:
AT5G35380, T26D22.11, T26D22_11
Chromosome Location:
Chr.5: 13593322 - 13596293 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02311365_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT5G35380
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_122929.2 NP_198388.1 7 - 8 970 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
protein kinase activity
ATP binding
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0