This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02312409_g1
See other SGR4 GE Assays ›
Gene Symbol:
SGR4
Gene Name:
Vesicle transport v-SNARE family protein
Gene Aliases:
AT5G39510, ATVTI11, ATVTI1A, IMPAIRED TONOPLAST TRAFFIC 3, ITT3, MUL8.190, MUL8_190, SHOOT GRAVITROPSIM 4, VESICLE TRANSPORT V-SNARE 11, VTI11, VTI1A, ZIG, ZIG1
Chromosome Location:
Chr.5: 15821293 - 15823840 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02312409_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123313.5 NP_198767.1 3 - 4 435 106
GenBank mRNA AY070486.1 - 3 - 4 285 106
AY091707.1 - 2 - 3 143 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
gravitropism
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
intracellular transport of virus
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0