This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02316556_g1
See other RAC2 GE Assays ›
Gene Symbol:
RAC2
Gene Name:
RAC-like 2
Gene Aliases:
ARABIDOPSIS THALIANA RAC 2, ARAC2, AT5G45970, ATRAC2, ATROP7, Arabidopsis RAC-like 2, MCL19.1, MCL19_1, RAC-like 2, RHO-RELATED PROTEIN FROM PLANTS 7, ROP7
Chromosome Location:
Chr.5: 18643540 - 18645976 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02316556_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_123965.2 NP_199409.1 4 - 5 585 77
GenBank mRNA U43026.1 - 4 - 5 391 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
establishment or maintenance of cell polarity
small GTPase mediated signal transduction
regulation of cell shape
actin cytoskeleton organization
cortical cytoskeleton organization
regulation of actin cytoskeleton organization
GTPase activity
GTP binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0