This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02317073_g1
See other VAM3 GE Assays ›
Gene Symbol:
VAM3
Gene Name:
Syntaxin/t-SNARE family protein
Gene Aliases:
ARABIDOPSIS THALIANA SYNTAXIN OF PLANTS 22, ARABIDOPSIS THALIANA VACUOLAR MORPHOLOGY 3, AT5G46860, ATSYP22, ATVAM3, SGR3, SHOOT GRAVITROPISM 3, SYNTAXIN OF PLANTS 22, SYP22, VACUOLAR MORPHOLOGY 3
Chromosome Location:
Chr.5: 19012021 - 19014110 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02317073_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124057.4 NP_568671.1 3 - 4 740 66
GenBank mRNA AF385695.1 - 3 - 4 575 66
AY072015.1 - 3 - 4 581 66
AY085121.1 - 3 - 4 597 66
AY133666.1 - 3 - 4 425 66
U88045.1 - 3 - 4 561 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle fusion
vacuole organization
amyloplast organization
negative gravitropism
stomatal movement
late endosome to vacuole transport
vesicle docking
membrane fusion
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline