This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02317074_g1
See other VAM3 GE Assays ›
Gene Symbol:
VAM3
Gene Name:
Syntaxin/t-SNARE family protein
Gene Aliases:
ARABIDOPSIS THALIANA SYNTAXIN OF PLANTS 22, ARABIDOPSIS THALIANA VACUOLAR MORPHOLOGY 3, AT5G46860, ATSYP22, ATVAM3, SGR3, SHOOT GRAVITROPISM 3, SYNTAXIN OF PLANTS 22, SYP22, VACUOLAR MORPHOLOGY 3
Chromosome Location:
Chr.5: 19012021 - 19014110 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02317074_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124057.4 NP_568671.1 4 - 5 797 71
GenBank mRNA AF385695.1 - 4 - 5 632 71
AY072015.1 - 4 - 5 638 71
AY085121.1 - 4 - 5 654 71
AY133666.1 - 4 - 5 482 71
U88045.1 - 4 - 5 618 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle fusion
vacuole organization
amyloplast organization
negative gravitropism
stomatal movement
late endosome to vacuole transport
vesicle docking
membrane fusion
SNARE binding
SNAP receptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0