This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02317347_g1
See other NUDT8 GE Assays ›
Gene Symbol:
NUDT8
Gene Name:
nudix hydrolase homolog 8
Gene Aliases:
AT5G47240, MQL5.10, MQL5_10, atnudt8, nudix hydrolase homolog 8
Chromosome Location:
Chr.5: 19183523 - 19185644 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02317347_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344713.1 NP_001331621.1 5 - 6 1018 150
NM_124095.4 NP_568680.1 5 - 6 970 150

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
response to wounding
NADH pyrophosphatase activity
ADP-ribose diphosphatase activity
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t6srt:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0