This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02317650_g1
See other NUDT2 GE Assays ›
Gene Symbol:
NUDT2
Gene Name:
nudix hydrolase homolog 2
Gene Aliases:
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 2, AT5G47650, ATNUDT2, ATNUDX2, MNJ7.24, MNJ7_24, nudix hydrolase homolog 2
Chromosome Location:
Chr.5: 19310192 - 19312747 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02317650_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001344736.1 NP_001331651.1 6 - 7 1051 87
NM_001344737.1 NP_001331652.1 7 - 8 732 87
NM_001344738.1 NP_001331649.1 6 - 7 678 87
NM_001344739.1 NP_001331650.1 6 - 7 611 87
NM_124139.4 NP_568687.1 7 - 8 782 87
GenBank mRNA AY042806.1 - 7 - 8 616 87
AY064646.1 - 6 - 7 540 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
signal transduction
NAD+ diphosphatase activity
NADH pyrophosphatase activity
ADP-ribose diphosphatase activity
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0