This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02319074_g1
See other CLC-C GE Assays ›
Gene Symbol:
CLC-C
Gene Name:
chloride channel C
Gene Aliases:
AT5G49890, ATCLC-C, CHLORIDE CHANNEL C, K9P8.3, K9P8_3, chloride channel C
Chromosome Location:
Chr.5: 20288055 - 20292432 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02319074_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLC-C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124367.4 NP_199800.1 2 - 3 703 88
GenBank mRNA AY120754.1 - 2 - 3 534 88
BT001211.1 - 2 - 3 414 88
Y09095.1 - 437 88
Z71447.1 - 2 - 3 447 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Process(es)

voltage-gated chloride channel activity
anion channel activity
protein binding
chloride transmembrane transporter activity
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline