This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02319301_g1
See other ELO3 GE Assays ›
Gene Symbol:
ELO3
Gene Name:
radical SAM domain-containing protein / GCN5-related N-acetyltransferase (GNAT) family protein
Gene Aliases:
AT5G50320, AtELP3, EAST1, ELONGATA 3, ELP3, Elongator protein 3, HAC8, HAG3, HISTONE ACETYLTRANSFERASE 8, HISTONE ACETYLTRANSFERASE OF THE GNAT FAMILY 3, MXI22.3, MXI22_3, enhancer-of-asymmetric leaves-two 1
Chromosome Location:
Chr.5: 20480626 - 20484945 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02319301_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELO3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124412.3 NP_568725.1 1 - 2 612 61
GenBank mRNA AJ964958.1 - 1 - 2 478 61
AY056323.1 - 1 - 2 506 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

tRNA wobble uridine modification
tRNA wobble base 5-methoxycarbonylmethyl-2-thiouridinylation
DNA mediated transformation
regulation of auxin mediated signaling pathway
organ growth
cell division
specification of plant organ axis polarity
regulation of leaf formation
histone acetyltransferase activity
N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline