This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02328231_g1
See other ATBET12 GE Assays ›
Gene Symbol:
ATBET12
Gene Name:
Target SNARE coiled-coil domain protein
Gene Aliases:
ARABIDOPSIS THALIANA BET1P/SFT1P-LIKE PROTEIN 14BB, AT4G14455, ATBS14B, BET12, BET1P/SFT1P-LIKE PROTEIN 14BB, BS14B
Chromosome Location:
Chr.4: 8310319 - 8312384 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02328231_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATBET12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_179053.3 NP_849384.1 5 - 6 524 119
GenBank mRNA AK117428.1 - 5 - 6 503 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
pollen tube growth
SNAP receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0