This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02352751_s1
See other ABCG18 GE Assays ›
Gene Symbol:
ABCG18
Gene Name:
ABC-2 type transporter family protein
Gene Aliases:
AT3G55110, ATP-binding cassette G18
Chromosome Location:
Chr.3: 20424550 - 20427384 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02352751_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_115369.4 NP_191071.1 2 1831 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transmembrane transport
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0