This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02354070_s1
See other ABCB24 GE Assays ›
Gene Symbol:
ABCB24
Gene Name:
ABC transporter of the mitochondrion 2
Gene Aliases:
ABC transporter of the mitochondrion 2, ARABIDOPSIS THALIANA ABC TRANSPORTER OF THE MITOCHONDRION 2, AT4G28620, ATATM2, ATM2, ATP-binding cassette B24, HALF-MOLECULE ABC TRANSPORTER ATM2, T5F17.70, T5F17_70
Chromosome Location:
Chr.4: 14135526 - 14137953 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02354070_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCB24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_119004.1 NP_194591.1 1 230 67
GenBank mRNA AF287698.1 - 1 230 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular iron ion homeostasis
response to cadmium ion
transmembrane transport
ATPase activity
transmembrane transporter activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline