This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt00516776_m1
See other UBE2K GE Assays ›
Gene Symbol:
UBE2K
Gene Name:
ubiquitin conjugating enzyme E2 K
Gene Aliases:
E2-25K, HIP2, HYPG, LIG
Chromosome Location:
Chr.6: 58774538 - 58854226 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt00516776_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174080.2 NP_776505.1 4 - 5 326 117
GenBank mRNA BC142324.1 - 4 - 5 489 117
S51016.1 - 4 - 5 326 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of peptidyl-threonine phosphorylation
free ubiquitin chain polymerization
regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to interferon-beta
positive regulation of type I interferon-mediated signaling pathway
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
ATP binding
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0