Assay Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034374.2 | NP_001029546.1 | 1 - 2 | 247 | 72 | |
| GenBank mRNA | BC102764.1 | - | 1 - 2 | 261 | 72 | |
| EE244269.1 | - | 1 - 2 | 248 | 72 | ||
More Information
Set Membership: |
|

MAPRE2
Genomic Map