Assay Details
| Gene Symbol: SSBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035478.1 | NP_001030555.1 | 8 - 9 | 607 | 93 | |
| GenBank mRNA | BC104606.1 | - | 8 - 9 | 607 | 93 | |
More Information
Set Membership: |
|

SSBP2
Genomic Map