This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt01076444_m1
See other PLEKHA8 GE Assays ›
Gene Symbol:
PLEKHA8
Gene Name:
pleckstrin homology domain containing A8
Gene Aliases:
FAPP-2
Chromosome Location:
Chr.4: 66219468 - 66328539 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt01076444_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLEKHA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197032.1 NP_001183961.1 9 - 10 1041 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
protein transport
ER to Golgi ceramide transport
glycolipid transport
ceramide 1-phosphate transport
glycolipid transporter activity
glycolipid binding
phosphatidylinositol-4-phosphate binding
ceramide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline