Assay Details
| Gene Symbol: AIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035018.2 | NP_001030190.2 | 5 - 6 | 685 | 81 | |
| GenBank mRNA | BC103049.1 | - | 5 - 6 | 686 | 81 | |
More Information
Set Membership: |
|

AIMP1
Genomic Map