This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt02660292_m1
See other MYDGF GE Assays ›
Gene Symbol:
MYDGF
Gene Name:
myeloid derived growth factor
Gene Aliases:
C19orf10, C7H19orf10
Chromosome Location:
Chr.7: 19470131 - 19482063 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt02660292_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYDGF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001164.1 NP_001001164.1 3 - 4 298 72
GenBank mRNA AY581200.1 - 3 - 4 298 72
BC102581.1 - 3 - 4 315 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s)

angiogenesis
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
apoptotic process
positive regulation of phosphatidylinositol 3-kinase signaling
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein kinase B signaling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline