This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03210588_m1
See other GTF2F1 GE Assays ›
Gene Symbol:
GTF2F1
Gene Name:
general transcription factor IIF subunit 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 18063616 - 18069587 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03210588_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015527.1 NP_001015527.1 2 - 3 257 80
GenBank mRNA BC146189.1 - 2 - 3 260 80
BT020716.1 - 2 - 3 257 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

response to virus
negative regulation of protein binding
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of catalytic activity
positive regulation of transcription from RNA polymerase II promoter
DNA binding
phosphatase activator activity
protein phosphatase binding
protein domain specific binding
promoter-specific chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0