This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03211282_m1
See other MOB3C GE Assays ›
Gene Symbol:
MOB3C
Gene Name:
MOB kinase activator 3C
Gene Aliases:
MOBKL2C
Chromosome Location:
Chr.3: 99514766 - 99523397 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03211282_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034755.1 NP_001029927.1 1 - 2 101 72
GenBank mRNA BC149870.1 - 1 - 2 121 72
BT020665.1 - 1 - 2 101 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC149870.1, NM_001034755.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0