This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03212693_m1
See other GLCE GE Assays ›
Gene Symbol:
GLCE
Gene Name:
glucuronic acid epimerase
Gene Aliases:
-
Chromosome Location:
Chr.10: 16029509 - 16139223 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03212693_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLCE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174070.2 NP_776495.1 2 - 3 824 81
GenBank mRNA AF003927.2 - 2 - 3 824 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
calcium ion binding
racemase and epimerase activity, acting on carbohydrates and derivatives
protein homodimerization activity
heparosan-N-sulfate-glucuronate 5-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0