This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03212975_m1
See other PPIB GE Assays ›
Gene Symbol:
PPIB
Gene Name:
peptidylprolyl isomerase B
Gene Aliases:
-
Chromosome Location:
Chr.10: 45818713 - 45824618 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03212975_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174152.2 NP_776577.1 2 - 3 240 108
GenBank mRNA BC102462.1 - 2 - 3 266 108
BT020966.1 - 2 - 3 276 108
D14073.1 - 2 - 3 240 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
positive regulation of multicellular organism growth
positive regulation by host of viral genome replication
protein stabilization
bone development
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
RNA polymerase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline