This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03213865_m1
See other GNGT2 GE Assays ›
Gene Symbol:
GNGT2
Gene Name:
G protein subunit gamma transducin 2
Gene Aliases:
-
Chromosome Location:
Chr.19: 37361994 - 37366300 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03213865_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNGT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174328.2 NP_776753.1 2 - 3 133 82
GenBank mRNA BC147970.1 - 2 - 3 387 82
U20085.1 - 2 - 3 133 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Process(es)

GTPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline