This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03216331_m1
See other GOT2 GE Assays ›
Gene Symbol:
GOT2
Gene Name:
glutamic-oxaloacetic transaminase 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 26447742 - 26471387 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03216331_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174806.2 NP_777231.1 8 - 9 1045 53
GenBank mRNA BC102303.1 - 8 - 9 1126 53
BT030486.1 - 8 - 9 1040 53
Z25466.1 - 1045 53

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
oxaloacetate metabolic process
aspartate metabolic process
aspartate biosynthetic process
glutamate metabolic process
fatty acid transport
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
response to ethanol
L-aspartate:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline