Assay Details
| Gene Symbol: STK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015602.1 | NP_001015602.1 | 7 - 8 | 866 | 58 | |
| GenBank mRNA | BC112753.1 | - | 7 - 8 | 871 | 58 | |
| BT020904.1 | - | 7 - 8 | 866 | 58 | ||
More Information
Set Membership: |
|

STK4
Genomic Map