This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03220847_m1
See other P2RX4 GE Assays ›
Gene Symbol:
P2RX4
Gene Name:
purinergic receptor P2X 4
Gene Aliases:
-
Chromosome Location:
Chr.17: 53925449 - 53941653 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03220847_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034049.1 NP_001029221.1 7 - 8 768 61
GenBank mRNA AF005155.1 - 3 - 4 251 61
BC120340.1 - 7 - 8 760 61
BT020829.1 - 7 - 8 768 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of blood pressure
positive regulation of calcium ion transport into cytosol
negative regulation of cardiac muscle hypertrophy
calcium-mediated signaling
ion transmembrane transport
response to fluid shear stress
purinergic nucleotide receptor signaling pathway
positive regulation of blood vessel endothelial cell migration
positive regulation of calcium-mediated signaling
membrane depolarization
regulation of cardiac muscle contraction
relaxation of cardiac muscle
excitatory postsynaptic potential
calcium ion transmembrane transport
cellular response to zinc ion
cellular response to ATP
apoptotic signaling pathway
positive regulation of endothelial cell chemotaxis
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding
identical protein binding
cadherin binding
ligand-gated calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0