This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03221643_m1
See other ATAT1 GE Assays ›
Gene Symbol:
ATAT1
Gene Name:
alpha tubulin acetyltransferase 1
Gene Aliases:
C23H6orf134, MEC17, TAT
Chromosome Location:
Chr.23: 28353032 - 28365839 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03221643_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276323.1 NP_001263252.1 12 - 13 1119 85
NR_075072.1 10 - 11 806 85
GenBank mRNA BM104818.1 - 4 - 5 410 85
BT021821.1 - 10 - 11 805 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron development
regulation of microtubule cytoskeleton organization
alpha-tubulin acetylation
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
tubulin N-acetyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0