Assay Details
| Gene Symbol: MS4A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077040.1 | NP_001070508.1 | 3 - 4 | 426 | 135 | |
| GenBank mRNA | BT021572.1 | - | 3 - 4 | 426 | 135 | |
More Information
Set Membership: |
|

MS4A7
Genomic Map