This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03223395_m1
See other FABP5 GE Assays ›
Gene Symbol:
FABP5
Gene Name:
fatty acid binding protein 5
Gene Aliases:
-
Chromosome Location:
Chr.14: 44488912 - 44494877 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03223395_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174315.3 NP_776740.1 1 - 2 129 82
GenBank mRNA AF059507.1 - 1 - 2 137 82
BC105196.1 - 1 - 2 147 82
BT020981.1 - 1 - 2 129 82
U55188.1 - 1 - 2 149 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
phosphatidylcholine biosynthetic process
negative regulation of glucose transport
regulation of prostaglandin biosynthetic process
positive regulation of peroxisome proliferator activated receptor signaling pathway
glucose homeostasis
regulation of sensory perception of pain
regulation of retrograde trans-synaptic signaling by endocanabinoid
positive regulation of cold-induced thermogenesis
retinoic acid binding
fatty acid binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-m5965:80/100.66.73.57:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0