Assay Details
| Gene Symbol: DHRS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174733.2 | NP_777158.1 | 3 - 4 | 778 | 119 | |
| GenBank mRNA | AF362071.1 | - | 3 - 4 | 778 | 119 | |
More Information
Set Membership: |
|

DHRS9
Genomic Map