This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03224498_m1
See other NDUFC2 GE Assays ›
Gene Symbol:
NDUFC2
Gene Name:
NADH:ubiquinone oxidoreductase subunit C2
Gene Aliases:
-
Chromosome Location:
Chr.29: 17979171 - 17987364 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03224498_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176642.3 NP_788815.1 2 - 3 467 107
GenBank mRNA BC102739.1 - 2 - 3 467 107
X68647.1 - 337 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0