This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03224639_m1
See other BRCA1 GE Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1 DNA repair associated
Gene Aliases:
-
Chromosome Location:
Chr.19: 43069050 - 43140130 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03224639_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178573.1 NP_848668.1 21 - 22 5445 56
GenBank mRNA AF479648.1 - 20 - 21 2043 56
AF479649.1 - 21 - 22 2154 56
AY077732.1 - 21 - 22 5445 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
regulation of transcription from RNA polymerase II promoter
fatty acid biosynthetic process
cell cycle
negative regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
protein autoubiquitination
protein K6-linked ubiquitination
DNA binding
transcription coactivator activity
ubiquitin-protein transferase activity
zinc ion binding
RNA polymerase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0