This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03224767_m1
See other GLUD1 GE Assays ›
Gene Symbol:
GLUD1
Gene Name:
glutamate dehydrogenase 1
Gene Aliases:
GDH, GDH 1, GDH1
Chromosome Location:
Chr.28: 41609012 - 41649016 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03224767_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLUD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182652.2 NP_872593.2 11 - 12 1620 63
GenBank mRNA AY138843.1 - 11 - 12 1383 63
AY256856.1 - 11 - 12 1508 63
BC103336.1 - 11 - 12 1570 63
EV666679.1 - 3 - 4 281 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glutamate catabolic process
glutamine metabolic process
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
ATP binding
GTP binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0