This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03224911_m1
See other CEACAM1 GE Assays ›
Gene Symbol:
CEACAM1
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 1
Gene Aliases:
CEACAM8
Chromosome Location:
Chr.18: 50817907 - 50837829 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03224911_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205788.1 NP_991357.1 6 - 7 1196 65
GenBank mRNA AY345127.1 - 6 - 7 1196 65
AY345128.1 - 6 - 7 1175 65
AY487416.1 - 6 - 7 1205 65
AY487417.1 - 6 - 7 1184 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline