This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03224989_g1
See other TPI1 GE Assays ›
Gene Symbol:
TPI1
Gene Name:
triosephosphate isomerase 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 103580104 - 103583461 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03224989_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TPI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013589.3 NP_001013607.1 4 - 5 486 64
GenBank mRNA BC102903.1 - 4 - 5 488 64
BT021064.1 - 4 - 5 468 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
methylglyoxal biosynthetic process
glyceraldehyde-3-phosphate biosynthetic process
triose-phosphate isomerase activity
methylglyoxal synthase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0