This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03226254_m1
See other HPGD GE Assays ›
Gene Symbol:
HPGD
Gene Name:
15-hydroxyprostaglandin dehydrogenase
Gene Aliases:
PGDH
Chromosome Location:
Chr.8: 6787672 - 6824508 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03226254_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HPGD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034419.2 NP_001029591.1 5 - 6 528 76
GenBank mRNA BC102458.1 - 5 - 6 529 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin metabolic process
transforming growth factor beta receptor signaling pathway
female pregnancy
parturition
ovulation
negative regulation of cell cycle
oxidation-reduction process
thrombin receptor signaling pathway
ductus arteriosus closure
regulation of prostaglandin catabolic process
prostaglandin E receptor activity
15-hydroxyprostaglandin dehydrogenase (NAD+) activity
NAD binding
NAD+ binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0