This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03226580_m1
See other NEIL3 GE Assays ›
Gene Symbol:
NEIL3
Gene Name:
nei like DNA glycosylase 3
Gene Aliases:
-
Chromosome Location:
Chr.27: 8574896 - 8621182 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03226580_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEIL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034490.2 NP_001029662.1 9 - 10 1701 71
GenBank mRNA BC105168.1 - 9 - 10 1702 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
nucleotide-excision repair
bubble DNA binding
damaged DNA binding
double-stranded DNA binding
single-stranded DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
zinc ion binding
DNA N-glycosylase activity
class I DNA-(apurinic or apyrimidinic site) endonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0