Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034612.2 | NP_001029784.1 | 1 - 2 | 95 | 66 | |
| GenBank mRNA | BC102687.1 | - | 1 - 2 | 96 | 66 | |
More Information
Set Membership: |
|

HPX
Genomic Map