This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03227304_m1
See other UNC119 GE Assays ›
Gene Symbol:
UNC119
Gene Name:
unc-119 lipid binding chaperone
Gene Aliases:
-
Chromosome Location:
Chr.19: 20009500 - 20015144 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03227304_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034645.2 NP_001029817.1 1 - 2 260 101
GenBank mRNA BC103449.1 - 1 - 2 261 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
endocytosis
lipoprotein transport
response to stimulus
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0