This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03227977_m1
See other TMIGD1 GE Assays ›
Gene Symbol:
TMIGD1
Gene Name:
transmembrane and immunoglobulin domain containing 1
Gene Aliases:
TMIGD
Chromosome Location:
Chr.19: 21399081 - 21466752 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03227977_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMIGD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035036.2 NP_001030208.1 2 - 3 382 80
GenBank mRNA BC102165.1 - 2 - 3 383 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Gene Ontology Categories:

Function(s)

regulation of cell migration
regulation of cell proliferation
negative regulation of apoptotic process
regulation of membrane permeability

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0