This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228390_m1
See other URI1 GE Assays ›
Gene Symbol:
URI1
Gene Name:
URI1 prefoldin like chaperone
Gene Aliases:
C18H19orf2, RMP
Chromosome Location:
Chr.18: 40542851 - 40608332 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228390_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: URI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035305.2 NP_001030382.1 7 - 8 978 75
GenBank mRNA BC107540.1 - 7 - 8 979 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of cell growth
regulation of transcription from RNA polymerase II promoter
response to virus
negative regulation of phosphatase activity
negative regulation of phosphoprotein phosphatase activity
cellular response to growth factor stimulus
cellular response to steroid hormone stimulus
negative regulation of intrinsic apoptotic signaling pathway
RNA polymerase II core binding
chromatin binding
transcription corepressor activity
protein phosphatase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0