This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228650_m1
See other H1-8 GE Assays ›
Gene Symbol:
H1-8
Gene Name:
H1.8 linker histone
Gene Aliases:
H1FOO
Chromosome Location:
Chr.22: 56187659 - 56223207 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228650_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H1-8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035372.1 NP_001030449.1 2 - 3 383 84
GenBank mRNA DQ206443.1 - 2 - 3 383 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
nucleosome positioning
histone exchange
regulation of DNA methylation
meiotic cell cycle
negative regulation of stem cell differentiation
nucleosomal DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0