This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03229739_m1
See other RBM11 GE Assays ›
Gene Symbol:
RBM11
Gene Name:
RNA binding motif protein 11
Gene Aliases:
-
Chromosome Location:
Chr.1: 22948041 - 22966832 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03229739_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040567.1 NP_001035657.1 3 - 4 352 105
GenBank mRNA BC102760.1 - 3 - 4 352 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0