This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03229747_m1
See other NUPR2 GE Assays ›
Gene Symbol:
NUPR2
Gene Name:
nuclear protein 2, transcriptional regulator
Gene Aliases:
NUPR1L
Chromosome Location:
Chr.25: 27771474 - 27772376 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03229747_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUPR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040574.1 NP_001035664.1 340 83
GenBank mRNA BC108184.1 - 340 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus

Gene Ontology Categories:

Function(s)

negative regulation of transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
cell cycle arrest
negative regulation of cell proliferation
cellular response to starvation
negative regulation of cell cycle arrest

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0