This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03229950_g1
See other ACADM GE Assays ›
Gene Symbol:
ACADM
Gene Name:
acyl-CoA dehydrogenase medium chain
Gene Aliases:
-
Chromosome Location:
Chr.3: 69086074 - 69124524 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03229950_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACADM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075235.1 NP_001068703.1 8 - 9 722 66
GenBank mRNA BC102989.1 - 8 - 9 722 66
EU009460.1 - 8 - 9 765 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

liver development
glycogen biosynthetic process
regulation of gluconeogenesis
fatty acid beta-oxidation
response to cold
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
cardiac muscle cell differentiation
acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0