This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03230443_m1
See other RBM4 GE Assays ›
Gene Symbol:
RBM4
Gene Name:
RNA binding motif protein 4
Gene Aliases:
-
Chromosome Location:
Chr.29: 44669757 - 44677607 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03230443_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077010.1 NP_001070478.1 3 - 4 1122 57
GenBank mRNA BC104570.1 - 3 - 4 1122 57
BT030489.1 - 3 - 4 1147 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
cap-independent translational initiation
IRES-dependent translational initiation
negative regulation of translation
cell differentiation
negative regulation of translation in response to stress
miRNA mediated inhibition of translation
entrainment of circadian clock by photoperiod
negative regulation of translational initiation
response to arsenic-containing substance
regulation of nucleocytoplasmic transport
positive regulation of muscle cell differentiation
circadian regulation of translation
RNA binding
mRNA binding
mRNA 3'-UTR binding
zinc ion binding
cyclin binding
miRNA binding
pre-mRNA binding
pre-mRNA intronic binding
pre-mRNA intronic pyrimidine-rich binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0